Q-Bio Taq Polymerase, MP Biomedicals

Supplier: MP Biomedicals
11EPQBT015 11EPQBT010 11EPQBT050 11EPQBT025 11EPQBT100
IC11EPQBT050EA 466.4 USD
IC11EPQBT050 IC11EPQBT015 IC11EPQBT025 IC11EPQBT100 IC11EPQBT010
Q-Bio Taq Polymerase, MP Biomedicals
For multiplex PCR and RAPDs.

  • Recombinant, truncated form of Thermus aquaticus
  • Highly thermostable (98 °C)
  • No 5 to 3' exonuclease activity
  • Enzyme cannot degrade the 5' end of primers
  • Store at –20 °C

Q-BioTaqTM DNA Polymerase is a thermostable DNA polymerase which can be used effectively for PCR and DNA sequencing by the chain termination method. This enzyme is encoded by a modified form of the Thermus aquaticus DNA polymerase gene with an N-terminal deletion. The properties of QBioTaqTM DNA Polymerase include high thermostability. This polymerase is particularly useful in applications where multiple amplifications are required. The enzyme is highly purified and free of RNases and endo- and exonucleases.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR