Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FastPROTEIN Red, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
116550700
Easy lysis of yeast cells to release recombinant proteins. The FastPROTEIN Red Kit is designed for rapid isolation and purification of protein from yeast cultures.

  • The process is designed to be a small scale test for protein induction
  • Instrument compatibility with FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-1
  • Protein Isolation from yeast, fungi and algae
  • DNase and RNase-free lysing matrix tubes

Easy lysis of yeast cells to release recombinant proteins The FastPROTEIN™ Red Matrix, also known as Lysing Matrix C, is used with the any FastPrep® Instrument to reproducibly lyse yeast cells within 40 seconds. A brief protocol is provided to allow for the maximum flexibility in lysis and purification methods. If no special induction buffer is required, the user can lyse yeast cells in the provided Yeast Breaking Buffer.

Rapid release of recombinant protein saves valuable time during optimization of expression. Information from up to 24 different induction variables can be quickly obtained after one 40 second FastPrep® run followed by electrophoresis. Once your protein of interest is expressed and identified, the active proteins released by the unique motion of the FastPrep® Instrument can be recovered from the lysate by a wide range of protein-specific techniques.

The FastPROTEINTM Red Matrix, also known as Lysing Matrix C, is used with the FastPrep®-24 or FastPrep® FP120 Instrument to reproducibly lyse yeast cells within 40 seconds. A brief protocol is provided to allow for the maximum fl exibility in lysis and purifi cation methods. If no special induction buffer is required, the user can lyse yeast cells in the provided yeast breaking buffer.

Application includes: Sample preparation; sample lysis; nucleic acid isolation and purification; molecular biology; gene expression; microbiological analysis; gene function; gene regulation.

Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ordering information: FastPrep® Instrument or alternative high speed disruptor, empty lysing matrix tubes and caps, microcentrifuge, pipettmen, purification reagents.




 

ORDER

Expand All / Collapse All
Description Size Includes Format Sample Type Supplier No. VWR Catalog Number Unit Price Quantity
Kit and Assay 100×2 ml Matrix C, 1.0 mm silica spheres Solution Feces; Yeast; Bacteria (Gram±); Fungi; Spores; Wastewater 116550700 IC116550700 Pack of 100 (200ml) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Storage Conditions Store at room temperature(15 - 30 °C)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty