Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Madrid Executive Mesh Vests, Class 2, Portwest

Supplier: PORTWEST LLC.
UC496YERXXXL UC496YERM UC496YERL UC496YERXXL UC496YERXL
Madrid Class 2 vest with dual ID holder, D-ring, and mic tab.

  • Cooling mesh upper torso, solid bottom
  • Front zipper closure
  • 100% polyester warp knit (solid portion), 3.5 oz.
  • 100% polyester mesh, 3 oz.
  • 2" silver reflective tape

These are designed with one cell phone, one pen and two front patch pockets with top and side entry.

Certifications: ANSI/ISEA 107-2015 Type R Class 2.



 

ORDER

Expand All / Collapse All
Clothing Size Material Color Supplier No. VWR Catalog Number Unit Price Quantity
M Polyester Yellow UC496YERM 76105-716 Each Retrieving
L Polyester Yellow UC496YERL 76105-714 Each Retrieving
XL Polyester Yellow UC496YERXL 76105-718 Each Retrieving
XXL Polyester Yellow UC496YERXXL 76105-720 Each Retrieving
3XL Polyester Yellow UC496YERXXXL 76105-722 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty