Tech-Med® Blood Pressure Kit, DUKAL Corporation®
Supplier: Dukal
2041
76236-420EA 63.02 USD
76236-420
Tech-Med® Blood Pressure Kit, DUKAL Corporation®
Blood Pressure Monitors
Blood pressure kit measures blood pressure from 0 to 300 mm Hg.
  • Black nylon cuff with index and range markings, and D-bar for self application
  • Oversized nylon carry case included
  • Manometer has a 10-year calibration warranty
Delivery information: Kit includes single head stethoscope, standard inflation system with two tube latex bladder, and vinyl carry case.