Gold Palladium (60:40), sputter coater target, Diameter 60 mm, Thickness 0.1 mm
Supplier: Electron Microscopy Sciences
91011
100492-482EA 1230.34 USD
100492-482
Gold Palladium (60:40), sputter coater target, Diameter 60 mm, Thickness 0.1 mm
Gold Palladium
These sputter coater targets are intended for EMS equipment and are stock items. Targets for other coaters are custom made and delivery is about one week. We are able to supply targets (foil with base) for the following sputtering units: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences and others.
  • Available in Gold, Gold/Palladium, Platnium, Chromium, and Silver
  • Up to 30% thicker than the original
Ordering information: When ordering, please state make and model of the sputtering unit to ensure proper fit.