    Sort by:
  • Search Within Results
Radotinib
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Radotinib is a BCR-ABL1-specific second-generation tyrosine kinase inhibitor (TKI).