    Sort by:
You Searched For:
GSK-269962
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   GSK-269962 is a potent Rho kinase (ROCK) inhibitor (IC₅₀ values are 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively)