    Sort by:
  • Search Within Results
WAY-170523
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   WAY-170523 is a potent and selective inhibitor of MMP-13 (IC₅₀ = 17 nM).