    Sort by:
  • Search Within Results
L-778123 hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   L-778123 hydrochloride is a potent inhibitor of FPTase and GGPTase-I with an IC₅₀ of 2 nM and 98 nM, respectively, in enzyme inhibition determination...