    Sort by:
You Searched For:
U-104
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   U-104 is a ureido-sulfonamide inhibitor of transmembrane carbonic anhydrase IX (CAIX) (Ki = 45.1 nM) and XII (CAXII) (Ki = 4.5 nM).