    Sort by:
  • Search Within Results
UNC0379
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   UNC0379 is a selective, substrate-competitive inhibitor of the lysine methyltransferase SET8 (IC₅₀ = 7.3 µM; Kd = 18.3 µM).