Would you like to log in again?
EPOWER, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0149E3
Experience the versatility of simply using one product for all of your QC needs, from media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing.
  • Available in concentrations ranging from 102 and 108 CFU per pellet
  • Strains are less than or equal to four passages from reference culture
  • Ready-to-use format to save you time and money
  • Refrigerated storage is easy and economical
  • Traceability to reference cultures ensures authenticity
  • Online Certificate of Analysis provides detailed strain information
  • Technical Support experts available for guidance
EPOWER is the embodiment of "flexibility is your friend". Use a single strain or combine multiple strains for a mixed microorganism population. Each quantitative microorganism pellet delivers a specific range of CFU, allowing users to easily manipulate this kit to deliver virtually any wide variety of concentration.