Dihydrorhodamine 123
Supplier: Anaspec Inc
Synonyms:
2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester
AS-85711
103011-336EA
171
USD
103011-336
Dihydrorhodamine 123
Dihydrorhodamine 123
Generic substrate for fluorimetric detection of oxidases (including peroxidase) in mitochondria
- Spectral properties: Abs/Em = 507/529 nm
- Solvent system: DMSO
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₂₁H₁₈N₂O₃ MW: 346.386 g/mol Storage Temperature: Freezer |
CAS Number:
109244-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...