2',7'-Dichlorofluorescein 3',6'-diacetate

Supplier: Anaspec Inc

Synonyms: H₂DCFDA2',7'-Dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester2',7'-Dichlorofluorescin diacetate

AS-85706
103011-330EA 171 USD
103011-330
2',7'-Dichlorofluorescein 3',6'-diacetate
2',7'-Dichlorofluorescein 3',6'-diacetate

Cell-permeable substrate for fluorimetric detection of oxidases (including peroxidase)


  • Spectral properties: Abs/Em = 495/529 nm
  • Solvent system: DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₂₄H₁₄Cl₂O₇
MW: 485.269 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037501
CAS Number: 2044-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR