D-(-)-Luciferin free acid, Ultrapure
Supplier: Anaspec Inc
Synonyms:
(S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
AS-82250
103011-092EA
160.77
USD
103011-092
D-(-)-Luciferin free acid, Ultrapure
D(-)-Luciferin
Luminescent substrate for firefly luciferase.
- Spectral properties: Abs/Em = 328/532 nm
- Solvent system: water
- Physical State: Solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₁₁H₈N₂O₃S₂ MW: 280.3228 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00042929 CAS Number: 2591-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...