4-Dimethylaminoazobenzene-4'-carboxylic acid

Supplier: Anaspec Inc

Synonyms: p-Methyl Red4-Dimethylaminoazobenzene-4-carboxylic AcidDABCYL acidPara Methyl Red

AS-81800
103011-046EA 144.69 USD
103011-046
4-Dimethylaminoazobenzene-4'-carboxylic acid
4-Dimethylaminoazobenzene-4'-carboxylic acid

DABCYL acid is the abbreviation of 4-(dimethylaminoazo)benzene-4-carboxylic acid. In some literature, DABSYL (4-dimethylaminoazobenzene-4'-sulfonyl chloride) is misused as ‘DABCYL’. DABCYL is one of the most popular acceptors for developing FRET-based nucleic acid probes and protease substrates. DABCYL dyes are often paired with EDANS in FRET-based fluorescent probes.


  • Spectral properties: Abs/Em = 425/none nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₁₅H₁₅N₃O₂
MW: 269.30 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058988
CAS Number: 6268-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR