5(6)-TAMRA cadaverine

Supplier: Anaspec Inc

Synonyms: Tetramethylrhodamine 5-(and-6)-carboxamide cadaverine

AS-81506
103011-028EA 384.38 USD
103011-028
5(6)-TAMRA cadaverine
5(6)-TAMRA cadaverine

5(6)-TAMRA cadaverine is a useful reagent for modifying carboxy groups via EDC-mediated reactions. It is a good glutamate transglutaminase substrate and a useful building block for small fluorescent molecules.


  • Spectral properties: Abs/Em = 544/570 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR