7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester ≥95% (by HPLC)

Supplier: Anaspec Inc

Synonyms: DNP-X acid, SE

AS-81211
103010-900EA 171 USD
103010-900
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester ≥95% (by HPLC)
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester

DEAC, SE is an excellent blue fluorescent building block for labeling amine-containing biomolecules.


  • Spectral properties: Abs/Em = 432/472 nm
  • Solvent system: DMF or Acetonitrile
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₁₈H₁₈N₂O₆
MW: 358.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467585
CAS Number: 139346-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR