6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

Supplier: Anaspec Inc

Synonyms: 6-TAMRA, SE6-TAMRA N-succinimidyl ester6-Carboxytetramethylrhodamine N-succinimidyl ester

AS-81126-
103010-852EA 238.23 USD
103010-852
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

6-TAMRA, SE is the other purified single isomer of 5(6)-TAMRA, SE. It is predominantly used for nucleotide labeling and DNA sequencing.


  • Spectral properties: Abs/Em = 547/573 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Storage Temperature: Freezer
CAS Number: 150810-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR