Magid® EconoWear Disposable Powder-Free Latex Finger Cots, Magid Glove
Supplier: Magid
T9776XL T9776M T9776S T9776L
13450-155PK 44.68 USD
13450-155 13450-149 13450-153 13450-151
Magid® EconoWear Disposable Powder-Free Latex Finger Cots, Magid Glove
Finger Cots Latex Finger Cots
These Econowear finger cots are a perfect for situations in which a full glove is not necessary.
  • Powder-free for reduced product contamination
  • Texturized tip for increased grip and sensitivity
  • 144 pieces per gross
This features a 7 cm (2.75"), 4 mil thickness for flexible protection, a rolled edge and a texturized tip for increased grip and sensitivity.
Caution: Product contains natural rubber latex which may cause allergic reactions in some users.