National Stereo Zoom Microscopes with Cameras
National®
BTW1-425T BTW2-425T
470308-140EA 1924.95 USD
470308-140 470308-142
National Stereo Zoom Microscopes with Cameras
Microscopes
Choose from 4 digital options.
  • On-board software for control and capture
  • Top and bottom LED illumination
  • Two 80 mm stage plates (frosted glass and reversible black/white plastic contrast plates)
Now with 4 digital options, the DC425 Series offers the convenience and versatility of viewing a large picture at 10× magnification, then zooming continuously all the way to close-up detail at 40× magnification. Upright, unreversed 3-dimensional image remains in focus throughout zoom range. Optional eyepieces and auxiliary objectives expand magnification range and working distances. Trinocular port accepts video or SLR adapters (included).