SP Bel-Art Aero Emergency Eye Wash Solution, Buffered pH Neutralizing, Stations and Bottles, Bel-Art Products, a part of SP
F24882-0005 F24882-0001 F24882-5016 F24882-0032 F24882-0016
76288-064PK 9.03 USD
76288-064 76288-066 76288-072 76288-070 76288-068
SP Bel-Art Aero Emergency Eye Wash Solution, Buffered pH Neutralizing, Stations and Bottles, Bel-Art Products, a part of SP
Immediate response eye first aid for the laboratory, classroom, work site, or public facilities.
  • Instant access to fast, safe and effective first aid for eye injuries caused by foreign particles such as dirt and debris, chemicals and other environmental irritants
  • 4 year shelf life from date of manufacture
  • Covered station (12W×13.25H×4.5"D) keeps bottles clean throughout shelf life
  • Carrying handle makes transport easy for field work
  • Wall mountable (hardware included)
  • Smaller 5 oz. and 0.5 oz. sizes can be easily included in portable workstations or in pockets for high risk situations
  • Quick twist top; easy to open and use
This multipurpose solution is effective in rinsing impurities and irritants from the eye, and in preventing damage from acids and corrosive substances that can occur after only 10 seconds. The pH neutralizing solution helps the eye return to a normal 7.4 pH level by quickly neutralizing corrosive chemicals.
All-in-one eye wash combines sterile saline with buffered pH neutralizing solution to help relieve irritation, discomfort and minimize likelihood of permanent damage. The unique eye-opener allows used fluids and contaminants to flow away from the eye (16 and 32 oz. bottles).
Certifications: Meets all FDA requirements for eye wash including buffered pH solution and preservative as per 21 CFR part 349. Meets ANSI Z358.1-2009 (as a personal eye wash only).