Anti-AOC3 Mouse Monoclonal Antibody [clone: TK8-14]

Supplier: Enzo Life Sciences
ABS657-0200
76285-174EA 589.97 USD
76285-174
Anti-AOC3 Mouse Monoclonal Antibody [clone: TK8-14]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, Flow Cytometry, IHC, IP, WB

Type: Primary
Antigen: AOC3
Clonality: Monoclonal
Clone: TK8-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR