Anti-RSV Human Monoclonal Antibody [clone: RSHZ19]

Supplier: Enzo Life Sciences
ABS655-0200
76285-170EA 589.97 USD
76285-170
Anti-RSV Human Monoclonal Antibody [clone: RSHZ19]
Antibodies
Recombinant monclonal antibody (Human IgG1κ).

Recommended Applications: ELISA

Type: Primary
Antigen: RSV
Clonality: Monoclonal
Clone: RSHZ19
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1κ
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR