Anti-p53 Mouse Monoclonal Antibody [clone: PAb421]

Supplier: Enzo Life Sciences
ABS653-0200
76285-166EA 615.79 USD
76285-166
Anti-p53 Mouse Monoclonal Antibody [clone: PAb421]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, IF, IHC (FS), IHC (PS), IP, WB

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: PAb421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human, Mouse, Rat, Rabbit, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR