Anti-NOTCH1 Mouse Monoclonal Antibody [clone: E6]

Supplier: Enzo Life Sciences
ABS647-0100
76285-154EA 589.97 USD
76285-154
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: E6]
Antibodies
Recombinant monclonal antibody (Mouse IgG1λ).

Recommended Applications: ELISA, IHC

Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1λ
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR