Anti-FN1 Human Monoclonal Antibody [clone: G4]

Supplier: Enzo Life Sciences
ABS635-0200
76285-130EA 589.97 USD
76285-130
Anti-FN1 Human Monoclonal Antibody [clone: G4]
Antibodies
Recombinant monclonal antibody (Human IgG1κ).

Recommended Applications: ELISA, IHC (FS), WB

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: G4
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR