Anti-FN1 Rabbit Monoclonal Antibody [clone: A2]

Supplier: Enzo Life Sciences
ABS634-0200
76285-128EA 589.97 USD
76285-128
Anti-FN1 Rabbit Monoclonal Antibody [clone: A2]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, IHC (FS), WB

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: A2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR