Anti-CD3E Rabbit Monoclonal Antibody [clone: YTH 12.5]

Supplier: Enzo Life Sciences
ABS624-0200
76284-610EA 600.38 USD
76284-610
Anti-CD3E Rabbit Monoclonal Antibody [clone: YTH 12.5]
Antibodies
Recombinant monclonal antibody (Rabbit IgGλ).

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: YTH 12.5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGλ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR