Anti-CD22 Human Monoclonal Antibody [clone: hL22]

Supplier: Enzo Life Sciences
ABS619-0200
76284-966EA 605.3 USD
76284-966
Anti-CD22 Human Monoclonal Antibody [clone: hL22]
Antibodies
Recombinant monclonal antibody (Human IgG1κ).

Recommended Applications: Flow Cytometry, IP, FUNC

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: hL22
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1κ
Reactivity: Human, Rhesus monkey, Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR