Anti-APP Mouse Monoclonal Antibody [clone: 6E10]

Supplier: Enzo Life Sciences
ABS611-0200
76284-950EA 600.38 USD
76284-950
Anti-APP Mouse Monoclonal Antibody [clone: 6E10]
Antibodies
Recombinant monclonal antibody (Mouse IgG1κ).

Recommended Applications: ELISA, IHC, IP, WB, Electron microscopy

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR