Anti-VEGFA Mouse Monoclonal Antibody [clone: A.4.6.1]

Supplier: Enzo Life Sciences
ABS603-0200
76285-114EA 589.97 USD
76285-114
Anti-VEGFA Mouse Monoclonal Antibody [clone: A.4.6.1]
Antibodies
Recombinant monclonal antibody (Mouse IgG1κ).

Recommended Applications: ELISA, Flow Cytometry, IHC, IP, WB, FUNC

Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: A.4.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR