Anti-V5 Tag Mouse Monoclonal Antibody [clone: SV5-P-K]

Supplier: Enzo Life Sciences
ABS594-0200
76285-096EA 589.97 USD
76285-096
Anti-V5 Tag Mouse Monoclonal Antibody [clone: SV5-P-K]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, IF, IHC (FS), IP, WB, Radioimmunoassay

Type: Primary
Antigen: V5 Tag
Clonality: Monoclonal
Clone: SV5-P-K
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR