Anti-TNF Human Monoclonal Antibody [clone: D2E7 (Adalimumab)]

Supplier: Enzo Life Sciences
ABS566-0200
76284-584EA 600.38 USD
76284-584
Anti-TNF Human Monoclonal Antibody [clone: D2E7 (Adalimumab)]
Antibodies
Recombinant monclonal antibody (Human IgG1κ).

Recommended Applications: ELISA, IF, IHC, FUNC

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: D2E7 (Adalimumab)
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR