Anti-IL2RA Rabbit Monoclonal Antibody [clone: Basiliximab]

Supplier: Enzo Life Sciences
ABS546-0200
76285-078EA 589.97 USD
76285-078
Anti-IL2RA Rabbit Monoclonal Antibody [clone: Basiliximab]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: Flow Cytometry, IF, IHC, WB, FUNC

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: Basiliximab
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR