Anti-IL2RA Rabbit Monoclonal Antibody [clone: Daclizumab]

Supplier: Enzo Life Sciences
ABS544-0200
76285-074EA 589.97 USD
76285-074
Anti-IL2RA Rabbit Monoclonal Antibody [clone: Daclizumab]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: Flow Cytometry, FUNC

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: Daclizumab
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR