Anti-CD38 Mouse Monoclonal Antibody [clone: HB7]

Supplier: Enzo Life Sciences
ABS438-0200
76285-016EA 589.97 USD
76285-016
Anti-CD38 Mouse Monoclonal Antibody [clone: HB7]
Antibodies
Recombinant monclonal antibody (Mouse IgG1κ).

Recommended Applications: Flow Cytometry, WB

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR