Anti-CEA Rabbit Monoclonal Antibody [clone: Arcitumomab]

Supplier: Enzo Life Sciences
ABS406-0200
76284-742EA 600.38 USD
76284-742
Anti-CEA Rabbit Monoclonal Antibody [clone: Arcitumomab]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: Flow Cytometry, IHC (PS)

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: Arcitumomab
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR