Human IgG3 kappa Isotype Control [clone: MOPC-21]

Supplier: Enzo Life Sciences
ABS583-0200
76284-918EA 589.97 USD
76284-918
Human IgG3 kappa Isotype Control [clone: MOPC-21]
Antibodies
Recombinant monclonal antibody (Human IgG3κ).

Recommended Applications: Flow Cytometry, WB

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG3kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR