Anti-IgG (H+L) Rabbit Antibody

Supplier: Southern Biotechnology
6026-01
103264-942EA 122.92 USD
103264-942
Anti-IgG (H+L) Rabbit Antibody
Antibodies
Purified Rabbit F(ab')2 Anti-Goat IgG(H+L) antibody with minimal reactivity to human serum proteins for use in ELISA, flow cytometry, and western blotting assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR