Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Anti-Sheep IgG (H+L) Antibody (HRP (Horseradish Peroxidase))

Supplier: SOUTHERN BIOTECHNOLOGY MS
6016-05
103264-936
Rabbit Anti-Sheep IgG (H+L) Antibody (HRP (Horseradish Peroxidase))
Rabbit F(ab')2 Anti-Sheep IgG(H+L)-HRP antibody with minimal reactivity to human serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty