Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti- IgA, IgG1, IgG2a, IgG2b, IgG3, IgM, Kappa, Lambda Antibody (HRP (Horseradish Peroxidase))

Supplier: SOUTHERN BIOTECHNOLOGY MS
5300-05
100244-906
Goat Anti- IgA, IgG1, IgG2a, IgG2b, IgG3, IgM, Kappa, Lambda Antibody (HRP (Horseradish Peroxidase))
The SBA Clonotyping System-HRP kit is designed for the isotyping of mouse monoclonal antibodies. It contains 2.5 mg of capture antibody and 1.0 mL of HRP conjugated anti-mouse Ig, mouse IgA, mouse IgG1, mouse IgG2a, mouse IgG2b, mouse IgG3, mouse IgM, mouse , mouse , and ABTS substrate. The kit may also be utilized for quantitative studies of mouse immunoglobulins in samples such as serum, supernatant, and ascites when used in conjunction with the Mouse Immunoglobulin Panel (SB Cat. No. 5300-01).

Type: Secondary
Antigen: IgA, IgG1, IgG2a, IgG2b, IgG3, IgM, Kappa, Lambda
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty