Anti-IgG Mouse Antibody (HRP (Horseradish Peroxidase)) [clone: SB108a]

Supplier: Southern Biotechnology
4700-05
103260-088EA 287.69 USD
103260-088
Anti-IgG Mouse Antibody (HRP (Horseradish Peroxidase)) [clone: SB108a]
Antibodies
HRP Anti-Monkey IgG for use in ELISA and western blotting assays.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: SB108a
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rhesus, Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR