Anti-IgG Mouse Antibody (Alexa Fluor® 555) [clone: 2A9]

Supplier: Southern Biotechnology
4090-32
103260-058EA 286.38 USD
103260-058
Anti-IgG Mouse Antibody (Alexa Fluor® 555) [clone: 2A9]
Antibodies
Alexa Fluor 555 Anti-Rabbit IgG antibody for use in immunohistochemistry and western blotting assays.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: 2A9
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR