Anti-Rabbit Ig Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB87a]

Supplier: Southern Biotechnology
4065-09
103260-056EA 309.92 USD
103260-056
Anti-Rabbit Ig Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB87a]
Antibodies
PE Anti-Rabbit Ig antibody for use in flow cytometry assays.

Type: Secondary
Antigen: Ig
Clonality: Monoclonal
Clone: SB87a
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR