Anti-Light Chain Mouse Antibody (HRP (Horseradish Peroxidase)) [clone: SB62a]

Supplier: Southern Biotechnology
4060-05
103260-054EA 396.23 USD
103260-054
Anti-Light Chain Mouse Antibody (HRP (Horseradish Peroxidase)) [clone: SB62a]
Antibodies
HRP Anti-Rabbit Light Chain antibody for use in ELISA and western blotting assays.

Type: Secondary
Antigen: Light Chain
Clonality: Monoclonal
Clone: SB62a
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR