Anti-IgG Goat Antibody

Supplier: Southern Biotechnology
4030-01
100244-750EA 100.69 USD
100244-750
Anti-IgG Goat Antibody
Antibodies
Purified Goat Anti-Rabbit IgG antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR