Anti-Rabbit IgM Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
4020-09
100244-748EA 281.15 USD
100244-748
Anti-Rabbit IgM Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Rabbit IgM-PE antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR