Anti-Rabbit IgM Goat Antibody

Supplier: Southern Biotechnology
4020-01
100244-736EA 100.69 USD
100244-736
Anti-Rabbit IgM Goat Antibody
Antibodies
Purified Goat Anti-Rabbit IgM antibody for use in ELISA, flow cytometry, immunohistochemistry, western blotting, ELISpot, and separation assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR