Anti-Kappa Mouse Antibody [clone: K4F5]

Supplier: Southern Biotechnology
3090-01
100244-638EA 367.46 USD
100244-638
Anti-Kappa Mouse Antibody [clone: K4F5]
Antibodies
Purified Anti-Rat Kappa antibody for use in flow cytometry, ELISA, immunohistochemistry, and western blotting assays.

Type: Secondary
Antigen: Kappa
Clonality: Monoclonal
Clone: K4F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR